missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPLAD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | PTPLAD1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274452
|
Novus Biologicals
NBP2-56963 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610457
|
Novus Biologicals
NBP2-56963-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTPLAD1 Polyclonal specifically detects PTPLAD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PTPLAD1 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 51495 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SIPIFNETGRFSFTLPYPVK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| B-ind1, BIND1, Butyrate-induced protein 1, butyrate-induced transcript 1, EC 4.2.1.-, FLJ90376, HACD3, hB-ind1, HSPC121,3-hydroxyacyl-CoA dehydratase 3, protein tyrosine phosphatase-like A domain containing 1, Protein tyrosine phosphatase-like protein PTPLAD1, Protein-tyrosine phosphatase-like A domain-containing protein 1 | |
| PTPLAD1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title