missing translation for 'onlineSavingsMsg'
Learn More

PTPIP51 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18415767 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415767 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415767 Supplier Novus Biologicals Supplier No. NBP147293V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTPIP51 Polyclonal antibody specifically detects PTPIP51 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPIP51
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Cerebral protein 10, FAM82Cmicrotubule-associated protein, family with sequence similarity 82, member A2, FLJ10579, hRMD-3, Protein FAM82A2, Protein FAM82C, Protein tyrosine phosphatase-interacting protein 51, ptpip51, PTPIP51family with sequence similarity 82, member C, regulator of microtubule dynamics 3, regulator of microtubule dynamics protein 3, RMD3, RMD-3, TCPTP-interacting protein 51
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 300 and 350 of human Protein Tyrosine Phosphatase-interacting Protein 51 (PTPIP51) using the numbering given in entry NP_060615.1 (GeneID 55177).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55177
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.