missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTP4A2 Monoclonal Antibody (5B6F4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531870
This item is not returnable.
View return policy
Description
MA5-31870 has been tested in indirect ELISA.
PRL-2, also known as Protein tyrosine phosphatase 4a2 (Ptp4a2) is a unique nuclear protein tyrosine phosphatase (PTP) that plays a central role in regulating diverse cellular processes. PRL-1 is induced in mitogen-stimulated cells and regenerating liver. PRL-2 exhibits 87% identity to PRL-1 in their amino acid sequences. All mouse PRL proteins contain a C-terminal consensus sequence for prenylation. All PRL proteins bear significant sequence homology to Cdc14p and the tumor suppressor PTEN/MMAC1. PRL-2 is preferentially expressed in skeletal muscle. PRL-2 is also expressed at lower levels in other tissues.
Specifications
| PTP4A2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q12974 | |
| PTP4A2 | |
| Purified recombinant fragment of human PTP4A2 (AA: 58-162) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| 5B6F4 | |
| Unconjugated | |
| PTP4A2 | |
| BM-008; HH13; HH7-2; HU-PP-1; OV-1; phosphatase of regenerating liver 2; PRL2; PRL-2; protein tyrosine phosphatase 4a2; protein tyrosine phosphatase IVA; protein tyrosine phosphatase IVA2; protein tyrosine phosphatase type IVA 2; protein tyrosine phosphatase type IVA, member 2; protein-tyrosine phosphatase 4a2; Protein-tyrosine phosphatase of regenerating liver 2; PTP(CAAXII); PTP4A; Ptp4a2; PTPCAAX2; ptp-IV1a; ptp-IV1b; tyrosine-phosphatase | |
| Mouse | |
| Protein G | |
| RUO | |
| 8073 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction