missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTP1B/PTPN2 Recombinant Rabbit Monoclonal Antibody (24GB1130)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | PTP1B/PTPN2 |
|---|---|
| Clone | 24GB1130 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276475
![]() |
Invitrogen™
MA556940 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274792
|
Invitrogen™
MA556941 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTP1B/PTPN2 Recombinant Monoclonal Antibody for Western Blot, Flow
PTP1B (Protein Tyrosine Phosphatase 1B), encoded by the PTPN1 gene located on chromosome 20q13.1, is a major regulator of several cellular signaling pathways, impacting diseases like diabetes, obesity, and various cancers. The protein structure includes multiple exons and functions as a non-transmembrane enzyme that negatively regulates signaling cascades, including insulin and leptin signaling, which are vital for metabolic homeostasis. PTPN2 (Protein Tyrosine Phosphatase Non-Receptor Type 2), located on chromosome 18p11.3-p11.2, also known as T-cell PTP, is similarly a non-transmembrane protein tyrosine phosphatase that modulates immune responses and cell growth. It has ten exons and produces two splice variants, impacting signaling pathways by regulating the phosphorylation state of key proteins. Both of these phosphatases are involved in the fine-tuning of signaling mechanisms that control cell growth, differentiation, and immune regulation, highlighting their importance in disease pathology and as potential therapeutic targets.Specifications
| PTP1B/PTPN2 | |
| 0.5 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17706, P18031, P20417, P35233, P35821, Q06180 | |
| 117063, 19246, 19255, 24697, 5770, 5771 | |
| A synthesized peptide derived from human PTP1B/PTPN2 (aa 11-66). | |
| Antibody |
| 24GB1130 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| PTPN1 | |
| protein tyrosine phosphatase 1B; PTP1B; T-cell protein tyrosine phosphatase; TCPTP | |
| PTPN1, PTPN2 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title