missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP1B/PTPN1 Rabbit anti-Human, Clone: 226, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89400-100UL
This item is not returnable.
View return policy
Description
PTP1B/PTPN1 Monoclonal specifically detects PTP1B/PTPN1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| PTP1B/PTPN1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 226 | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate | |
| EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 | |
| Recombinant Human PTP1B/PTPN1 protein | |
| 100 μg | |
| Phospho Specific | |
| 5770 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction