missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP1B/PTPN1 Antibody (OTI1D10), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
PTP1B/PTPN1 Monoclonal antibody specifically detects PTP1B/PTPN1 in Human, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | PTP1B/PTPN1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI1D10 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human PTPN1(NP_002818) produced in HEK293 cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?