missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PTP1B/PTPN1 Monoclonal antibody specifically detects PTP1B/PTPN1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Antigen | PTP1B/PTPN1 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 226 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot, ELISA, Immunoprecipitation |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human PTP1B/PTPN1 (Accession#: NP_002818.1; Glu 2-Asn 321). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?