missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP1B/PTPN1 Antibody (226), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP2-89400AF700
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
PTP1B/PTPN1 Monoclonal specifically detects PTP1B/PTPN1 in Human samples. It is validated for Western Blot, ELISA.
Specifica
| PTP1B/PTPN1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PTP1B/PTPN1 (NP_002818.1; Glu 2-Asn 321). | |
| 0.1 mL | |
| Phospho Specific | |
| 5770 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 226 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto