missing translation for 'onlineSavingsMsg'
Learn More

PTP1B/PTPN1 Antibody (226), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18384841 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384841 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384841 Supplier Novus Biologicals Supplier No. NBP289400AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PTP1B/PTPN1 Monoclonal specifically detects PTP1B/PTPN1 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP1B/PTPN1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 226
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PTP1B/PTPN1 (NP_002818.1; Glu 2-Asn 321).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5770
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.