missing translation for 'onlineSavingsMsg'
Learn More

PTP1B-IN-2, MedChemExpress

Product Code. p-200099767
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493028

Brand: Medchem Express HY100462

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PTP1B-IN-2 is a potent protein tyrosine phosphatase 1B (PTP1B) inhibitor with an IC50 of 50 nM.

PTP1B-IN-2 is a potent protein tyrosine phosphatase 1B (PTP1B) inhibitor with an IC50 of 50 nM.

Specifications

Chemical Name or Material PTP1B-IN-2
CAS 1919853-46-5
Color White
Purity Grade Notes Research
Molecular Formula C34H36N2O9S2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (146.89 mM)
SMILES O=S(N(C1=CC(C(OC)=O)=C(OCC2=CC=C(C)C=C2)C=C1)CC3=CC=C(N(S(C)(=O)=O)CC(OC)=O)C=C3)(CC4=CC=CC=C4)=O
Molecular Weight (g/mol) 680.79
Formula Weight 680.79
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.