missing translation for 'onlineSavingsMsg'
Learn More

PTP1B-IN-1, MedChemExpress

Product Code. 16495066
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495066 100 mg 100mg
16474098 10 mg 10mg
16450457 50 mg 50mg
16406807 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495066 Supplier Medchem Express Supplier No. HY10704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PTP1B-IN-1 is a potent protein tyrosine phosphatase-1B (PTP1B) inhibitor with IC50 of 1.6 mM; 1,2,5-thiadiazolidin-3-one-1,1-dioxide scaffold for derivatives synthesis.

PTP1B-IN-1 is a potent protein tyrosine phosphatase-1B (PTP1B) inhibitor with IC50 of 1.6 mM

Specifications

Chemical Name or Material PTP1B-IN-1
Physical Form Solid
CAS 612530-44-6
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H8N2O3S
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym PTP1B inhibitor
Solubility Information DMSO : ≥ 38 mg/mL (179.05 mM)
SMILES O=S(N1C2=CC=CC=C2)(NC(C1)=O)=O
Molecular Weight (g/mol) 212.23
Formula Weight 212.23
Percent Purity 98.84%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.