missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP sigma/PTPRS Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92999-0.02ml
This item is not returnable.
View return policy
Description
PTP sigma/PTPRS Polyclonal antibody specifically detects PTP sigma/PTPRS in Rat samples. It is validated for Western Blot
Specifications
| PTP sigma/PTPRS | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.1.3.48, protein tyrosine phosphatase PTPsigma, protein tyrosine phosphatase, receptor type, S, protein tyrosine phosphatase, receptor type, sigma, PTPSIGMA, receptor-type tyrosine-protein phosphatase S, Receptor-type tyrosine-protein phosphatase sigma, R-PTP-S, R-PTP-sigma | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1100-1280 of human PTPRS (NP_002841.3). TNRGSSLGGLQQTVTAWTAFNLLNGKPSVAPKPDADGFIMVYLPDGQSPVPVQSYFIVMVPLRKSRGGQFLTPLGSPEDMDLEELIQDISRLQRRSLRHSRQLEVPRPYIAARFSVLPPTFHPGDQKQYGGFDNRGLEPGHRYVLFVLAVLQKSEPTFAASPFSDPFQLDNPDPQPIVDGE | |
| 0.02 mL | |
| Primary | |
| Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5802 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction