missing translation for 'onlineSavingsMsg'
Learn More

PTP Sample for Sulfur in Crude Oil Analysis

Product Code. 30312835
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 30312835

Brand: VHG Labs VHGPTPSCRD25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium∣Antimony∣Barium∣Boron∣Cadmium∣Calcium∣Chromium∣Copper∣Iron∣Lead∣Magnesium∣Manganese∣Molybdenum∣Nickel∣Phosphorus∣Potassium∣Silver∣Sodium∣Sulfur∣Tin∣Vanadium∣Zinc
CAS 7429-90-5, 7440-36-0, 7440-39-3, 7440-42-8, 7440-43-9, 7440-70-2
Analyte or Component Names Aluminium∣Antimony∣Barium∣Boron∣Cadmium∣Calcium∣Chromium∣Copper∣Iron∣Lead∣Magnesium∣Manganese∣Molybdenum∣Nickel∣Phosphorus∣Potassium∣Silver∣Sodium∣Sulfur∣Tin∣Vanadium∣Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO/IEC 17043
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Single Solution
Element Multi-element
Solvent or Matrix Crude Oil
Industry Type Petrochemical Testing
Quantity 25 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.