missing translation for 'onlineSavingsMsg'
Learn More

PTP rho/PTPRT Antibody [Allophycocyanin], Novus Biologicals™

Product Code. 30647448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30647448 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30647448 Supplier Novus Biologicals Supplier No. AF3697A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

PTP rho/PTPRT Polyclonal antibody specifically detects PTP rho/PTPRT in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP rho/PTPRT
Applications Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias EC 3.1.3, EC 3.1.3.48, KIAA0283RPTPrho, protein tyrosine phosphatase, receptor type, T, receptor protein tyrosine phosphatase, Receptor-type tyrosine-protein phosphatase rho, receptor-type tyrosine-protein phosphatase T, RPTP-rho, R-PTP-T
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human PTP rho /PTPRT, Gln26-Lys725, Accession # O14522
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 11122
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.