missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP pi/PTPRU Antibody (PTPRU/7616), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23865-20ug
This item is not returnable.
View return policy
Description
PTP pi/PTPRU Monoclonal antibody specifically detects PTP pi/PTPRU in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PTP pi/PTPRU | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| FMI, hPTP-J, pancreatic carcinoma phosphatase 2, PCP2, PCP-2, pi R-PTP-Psi, protein tyrosine phosphatase J, protein tyrosine phosphatase, receptor type, U, protein-tyrosine phosphatase J, protein-tyrosine phosphatase pi, PTP, PTP pi, PTP-J, PTP-PI, PTPPSI, PTP-RO, Receptor protein tyrosine phosphatase hPTP-J, R-PTP-PSI | |
| Recombinant fragment (around aa1-200) of human PTP pi/PTPRU protein (exact sequence is proprietary) | |
| 20 μg | |
| Signal Transduction | |
| 10076 | |
| Store at 4°C. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| PTPRU/7616 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction