missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP mu/PTPRM Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-39013
This item is not returnable.
View return policy
Description
PTP mu/PTPRM Polyclonal antibody specifically detects PTP mu/PTPRM in Human, Mouse, Rat, Equine, Hamster, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PTP mu/PTPRM | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| EC 3.1.3.48, hR-PTPu, MGC166994, protein tyrosine phosphatase mu, protein tyrosine phosphatase, receptor type, M, protein tyrosine phosphatase, receptor type, mu polypeptide, Protein-tyrosine phosphatase mu, PTPRL1R-PTP-MU, receptor-type tyrosine-protein phosphatase mu, RPTPM, R-PTP-mu, RPTPU | |
| Synthetic 18 amino acid peptide from internal region of human PPTP mu/PTPRM. | |
| 0.05 mg | |
| Protein Phosphatase | |
| 5797 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 15 μg/mL | |
| P28827 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Equine, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction