missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP epsilon Antibody (OTI5F2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73713H
This item is not returnable.
View return policy
Description
PTP epsilon Monoclonal specifically detects PTP epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PTP epsilon | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| PTPRE | |
| Full length recombinant protein of human PTP epsilon (NP_006495) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5F2 | |
| Western Blot, Immunohistochemistry | |
| DKFZp313F1310, EC 3.1.3.48, FLJ57799, FLJ58245, HPTPE, protein tyrosine phosphatase epsilon, protein tyrosine phosphatase, receptor type, E, protein tyrosine phosphatase, receptor type, epsilon polypeptide, Protein-tyrosine phosphatase epsilon, PTPE, receptor-type tyrosine-protein phosphatase epsilon, R-PTP-EPSILON | |
| Mouse | |
| Affinity Purified | |
| GPCR, Protein Phosphatase, Tyrosine Kinases | |
| 5791.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction