missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP epsilon Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87189
This item is not returnable.
View return policy
Description
PTP epsilon Polyclonal specifically detects PTP epsilon in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PTP epsilon | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| DKFZp313F1310, EC 3.1.3.48, FLJ57799, FLJ58245, HPTPE, protein tyrosine phosphatase epsilon, protein tyrosine phosphatase, receptor type, E, protein tyrosine phosphatase, receptor type, epsilon polypeptide, Protein-tyrosine phosphatase epsilon, PTPE, receptor-type tyrosine-protein phosphatase epsilon, R-PTP-EPSILON | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PTPRE | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NGILEEQEQQRVMLLSRSPSGPKKYFPIPVEHLEEEIRIRSADDCKQFREEFNSLPSGHIQGTFELANKEENREKNRYPNILPNDHSRVILSQLDGIPCSDYINASYIDGYKEKNKFIAAQGPKQE | |
| 0.1 mL | |
| GPCR, Protein Phosphatase, Tyrosine Kinases | |
| 5791 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction