Learn More
Invitrogen™ PTOV1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA594928
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Kidney Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
PTOV1 may activate transcription. Required for nuclear translocation of FLOT1. Promotes cell proliferation.
Specifications
| PTOV1 | |
| Polyclonal | |
| Unconjugated | |
| PTOV1 | |
| 1110030G05Rik; ACID2; Activator interaction domain-containing protein 2; AU041779; Gcap3; granule cell antiserum positive 3; PP642; prostate tumor over expressed gene 1; prostate tumor overexpressed 1; prostate tumor-overexpressed gene 1 protein; Prostate tumor-overexpressed gene 1 protein homolog; Ptov1; PTOV-1; UNQ6127/PRO20092 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 292888, 53635 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| Antibody with 5mg BSA, 0.2mg Na2PO4, 0.9mg NaCl and 0.05mg sodium azide | |
| Q5U2W6, Q86YD1 | |
| PTOV1 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human PTOV1 (385-414aa NFVNGIRRVIANQQQVLQRNLEQEQQQRGM). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.