missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTMA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561459
This item is not returnable.
View return policy
Description
Immunogen sequence: EVVEEAENGR DAPANGNANE ENGEQEADNE VDEEEE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
The thymus gland produces several hormones or hormone-like substances which are derived from a polypeptide precursor containing (in the rat) 113 amino acids and known as prothymosin-alpha. A peptide containing 28 amino acid residues, named thymosin-alpha-1, was originally isolated from calf thymosin fraction 5 and shown to restore various aspects of immune function in several in vitro and invivo test systems. Thymosin-alpha-1 was subsequently isolated from a similar fraction from human thymosin and reported to have the same amino acid sequence as bovine thymosin-alpha-1. In 2003, Jiang et al. identified a pathway that regulates mitochondria-initiated caspase activity. In this pathway, putative human HLA class II-associated protein (PHAP) promoted caspase-9 activation after apoptosome formation, whereas prothymosin-alpha-1 negatively regulated caspase-9 activation by inhibiting apoptosome formation.
Specifications
| PTMA | |
| Polyclonal | |
| Unconjugated | |
| PTMA | |
| alpha-prothymosin; gene sequence 28; prothymosin alpha; prothymosin alpha protein; Prothymosin alpha, N-terminally processed; prothymosin, alpha; prothymosin-alpha; PTMA; Thym; thymosin; Thymosin alpha; Thymosin alpha-1; TMSA; TMSB4; TMSB4X | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5757 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P06454 | |
| PTMA | |
| Recombinant protein corresponding to Human PTMA. Recombinant protein control fragment (Product #RP-99547). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction