missing translation for 'onlineSavingsMsg'
Learn More

PTK9 Antibody (5C4), Novus Biologicals™

Product Code. 18326848 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326848 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326848 Supplier Novus Biologicals Supplier No. H00005756M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTK9 Monoclonal antibody specifically detects PTK9 in Human, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTK9
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5C4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH22344
Gene Alias A6, A6 protein tyrosine kinase, Protein A6, Protein tyrosine kinase 9MGC23788, PTK9 protein tyrosine kinase 9, PTK9MGC41876, twinfilin, actin-binding protein, homolog 1 (Drosophila), twinfilin-1
Host Species Mouse
Immunogen PTK9 (AAH22344, 80 a.a. ~ 252 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSSAKPKGPAGKRGIRRLIRGPAETEATTD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5756
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.