missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTK7/CCK4 Antibody (525222) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4499JF525
This item is not returnable.
View return policy
Description
PTK7/CCK4 Monoclonal antibody specifically detects PTK7/CCK4 in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot,Simple Western
Specifications
| PTK7/CCK4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7 | |
| E. coli-derived recombinant human PTK7/CCK4, Ala31-Ser199, Accession # Q13308 | |
| RUO | |
| Primary | |
| Human, Rat | |
| Liquid |
| Immunohistochemistry, Western Blot, Simple Western | |
| 525222 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Protein Kinase | |
| 5754 | |
| Store at 4°C in the dark. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu