missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTHLH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102455
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PTHLH.
The protein encoded by this gene is a member of the parathyroid hormone family. This hormone regulates endochondral bone development and epithelial-mesenchymal interactions during the formation of the mammary glands and teeth. This hormone is involved in lactation possibly by regulating the mobilization and transfer of calcium to the milk. The receptor of this hormone, PTHR1, is responsible for most cases of humoral hypercalcemia of malignancy. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed. There is also evidence for alternative translation initiation from non-AUG (CUG and GUG) start sites, in-frame and downstream of the initiator AUG codon, to give rise to nuclear forms of this hormone.
Specifications
| PTHLH | |
| Polyclonal | |
| Unconjugated | |
| PTHLH | |
| BDE2; calciotropic hormone, growth factor; HHM; osteostatin; parathyroid hormone like hormone; parathyroid hormone related peptide; parathyroid hormone-like hormone; parathyroid hormone-like hormone preproprotein; parathyroid hormone-like peptide; parathyroid hormone-like protein; parathyroid hormone-like related protein; parathyroid hormone-related peptide; parathyroid hormone-related protein; parathyroid-like peptide; PLP; PTHLH; PTH-like; PTHR; PTH-related peptide; PTH-related protein; Pthrp; PTH-rP; PTHrP[107-139]; PTHrP[1-36]; PTHrP[38-94] | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19227, 24695, 5744 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P12272, P13085, P22858 | |
| PTHLH | |
| A synthesized peptide derived from human PTHLH(Accession P12272), corresponding to amino acid residues T75-K125. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction