missing translation for 'onlineSavingsMsg'
Learn More

PTH1R/PTHR1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18356033 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356033 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356033 Supplier Novus Biologicals Supplier No. NBP299962JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTH1R/PTHR1 Polyclonal specifically detects PTH1R/PTHR1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTH1R/PTHR1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias MGC138426, parathyroid hormone 1 receptorMGC138452, parathyroid hormone/parathyroid hormone-related peptide receptor, parathyroid hormone/parathyroid hormone-related protein receptor, PFE, PTH/PTHr receptor, PTH/PTHrP type I receptor, PTH1 receptor, PTHRPTHR1parathyroid hormone receptor 1, seven transmembrane helix receptor
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PTH1R/PTHR1 (Q03431; Met 1-Gly 188).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5745
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.