missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH1R/PTHR1 Antibody (1G7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26359-100ul
This item is not returnable.
View return policy
Description
PTH1R/PTHR1 Monoclonal antibody specifically detects PTH1R/PTHR1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| PTH1R/PTHR1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1G7 | |
| Western Blot 1:500-1:5000, ELISA | |
| MGC138426, parathyroid hormone 1 receptorMGC138452, parathyroid hormone/parathyroid hormone-related peptide receptor, parathyroid hormone/parathyroid hormone-related protein receptor, PFE, PTH/PTHr receptor, PTH/PTHrP type I receptor, PTH1 receptor, PTHRPTHR1parathyroid hormone receptor 1, seven transmembrane helix receptor | |
| A synthesized peptide derived from Human PTH1R/PTHR1 [UniProt Q03431] | |
| 100 μL | |
| Biologically Active Proteins, GPCR, Neuroscience, Signal Transduction | |
| 5745 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction