missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH1R Polyclonal Antibody

PTH1R Polyclonal Antibody
Brand: Invitrogen PA580955
This item is not returnable.
View return policy
Description
PTH1R Polyclonal Antibody for IHC (P), ELISA
Specifications
| PTH1R | |
| Polyclonal | |
| Unconjugated | |
| Pth1r | |
| MGC138426; MGC138452; Parathormone; Parathyrin; parathyroid hormone 1 receptor; parathyroid hormone receptor; parathyroid hormone receptor 1; parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PFE; PPR; PTH; PTH/PTHr receptor; PTH/PTHrP receptor; PTH/PTHrP type I receptor; PTH1 receptor; PTH1R; Pthr; Pthr1; PTHrel; PTH-related peptide receptor; PTHrP-R; PTHrP-receptor; seven transmembrane helix receptor | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 5745 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q03431 | |
| Pth1r | |
| Recombinant Human PTH1R/PTHR1 protein (Met1-Gly188). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction