missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH, Rabbit anti-Human, Clone: PTH/2295R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-79759-100UG
This item is not returnable.
View return policy
Description
PTH Monoclonal specifically detects PTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| PTH | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P01270, P01270 | |
| PTH | |
| A recombinant fragment around aa 32-115 of human mature-PTH-polypeptide (exact sequence is proprietary) | |
| 100 μg | |
| Apoptosis, Biologically Active Proteins, Cancer | |
| 5741 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| PTH/2295R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Epitope of this monoclonal antibody maps in between aa 1-34 of PTH. | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction