missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH Rabbit anti-Human, Clone-PTH/1717R, Novus Biologicals™
Rabbit Monoclonal Antibody
£473.00
Specifications
| Antigen | PTH |
|---|---|
| Clone | PTH/1717R |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Description
PTH Monoclonal specifically detects PTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PTH | |
| 1 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 | |
| PTH | |
| IgG κ | |
| Protein A or G purified | |
| 9 kDa |
| PTH/1717R | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins, Cancer | |
| PBS with No Preservative | |
| 5741 | |
| Recombinant human PTH protein N-terminal fragment (exact sequence is proprietary) | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title