missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTH Monoclonal Antibody (BGN/1F8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA183385
This item is not returnable.
View return policy
Description
Mouse anti Human parathyroid hormone monoclonal antibody, clone BGN/1F8 recognizes human parathyroid hormone (PTH) also known as Parathyrin.
This gene encodes a member of the parathyroid family of proteins. The encoded preproprotein is proteolytically processed to generate a protein that binds to the parathyroid hormone/parathyroid hormone-related peptide receptor and regulates blood calcium and phosphate levels. Excess production of the encoded protein, known as hyperparathyroidism, can result in hypercalcemia and kidney stones. On the other hand, defective processing of the encoded protein may lead to hypoparathyroidism, which can result in hypocalcemia and numbness.
Specifications
| PTH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P01270, P04089 | |
| PTH | |
| Synthetic peptide corresponding to amino acids 1-34 of mature PTH conjugated to a a proprietary carrier molecule. | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| BGN/1F8 | |
| Unconjugated | |
| PTH | |
| hypothalamic parathyroid hormone; parathormone; Parathyrin; parathyroid hormone; parathyroid hormone 1; preproparathyroid hormone; prepro-PTH; PTH; PTH-(1-84); Pth1; Pthp; Pthr1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 19226, 24694, 5741 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction