missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTGIS Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA553132

Product Code. 15711081

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LTLYGIEALP RTHESQAQDR VHSADVFHTF RQLDRLLPKL ARGSLSVGDK DHMCSVKSRL WKLLSPARLA RRAHRSKWLE SYLLHLEEMG VSEEMQARAL VLQLWATQGN MGPAAFWLLL FLLKNPEALA AVRGELESIL WQAEQ Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 85%.

CYP8A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. However, this protein is considered a member of the cytochrome P450 superfamily on the basis of sequence similarity rather than functional similarity. This endoplasmic reticulum membrane protein catalyzes the conversion of prostglandin H2 to prostacyclin (prostaglandin I2), a potent vasodilator and inhibitor of platelet aggregation. An imbalance of prostacyclin and its physiological antagonist thromboxane A2 contribute to the development of myocardial infarction, stroke, and atherosclerosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTGIS
Polyclonal
Unconjugated
PTGIS
Cyp8; CYP8A1; cytochrome P450 8A1; cytochrome P450, family 8, subfamily A, polypeptide 1; hydroperoxy icosatetraenoate dehydratase; LOW QUALITY PROTEIN: prostacyclin synthase; MGC126858; MGC126860; PGIS; Prostacyclin synthase; prostaglandin I2 (prostacyclin) synthase; prostaglandin I2 (prostacyclin) synthase like; prostaglandin I2 synthase; PTGI; ptgis; ptgisl
Rabbit
Antigen affinity chromatography
RUO
5740
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q16647
PTGIS
Recombinant protein corresponding to Human PTGIS. Recombinant protein control fragment (Product #RP-90174).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.