missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PTGIR Antibody (2A7), Novus Biologicals™

Product Code. 18346598 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346598 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346598 Supplier Novus Biologicals™ Supplier No. H00005739M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTGIR Monoclonal antibody specifically detects PTGIR in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGIR
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 2A7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000951
Gene Alias IP, MGC102830, PGI receptor, PGI2 receptor, PRIPR, prostacyclin receptor, prostaglandin I2 (prostacyclin) receptor (IP), Prostaglandin I2 receptor, Prostanoid IP receptor
Host Species Mouse
Immunogen PTGIR (NP_000951, 296 a.a. ∽ 386 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RKAVFQRLKLWVCCLCLGPAHGDSQTPLSQLASGRRDPRAPSAPVGKEGSCVPLSAWGEGQVEPLPPTQQSSGSAVGTSSKAEASVACSLC
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 5739
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.