missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTGFR Monoclonal Antibody (1014602)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA560876

Product Code. 30535775

  • £357.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene is member of the G-protein coupled receptor family. This protein is a receptor for prostaglandin F2-alpha (PGF2-alpha), which is known to be a potent luteolytic agent, and may also be involved in modulating intraocular pressure and smooth muscle contraction in uterus. Knockout studies in mice suggest that the interaction of PGF2-alpha with this receptor may initiate parturition in ovarian luteal cells and thus induce luteolysis. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTGFR
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P43088
PTGFR
Synthetic peptide containing human PTGFR RD3 epitope.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry
1014602
Unconjugated
PTGFR
AI957154; fp; FP prostanoid receptor; FP prostanoid receptor FPB isoform; FP receptor; PF2AR; PGF; PGF receptor; PGF2 alpha receptor; PGF2-alpha receptor; prostaglandin F receptor; prostaglandin F receptor (FP); prostaglandin F2 alpha receptor; prostaglandin F2alpha receptor; prostaglandin F2-alpha receptor; prostaglandin F2alpha receptor FPa isoform; prostaglandin F2alpha receptor isoform-aplpha; prostaglandin F2alpha receptor isoform-beta; prostaglandin F2alpha receptor isoform-delta; prostaglandin F2alpha receptor isoform-epsiron; prostaglandin F2alpha receptor isoform-gamma; prostaglandin receptor (2-alpha); prostanoid FP receptor; Ptgfr
Mouse
Protein A/G
RUO
5737
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.