missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGER3, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB31423.100uL
This item is not returnable.
View return policy
Description
Sequence: MKETRGYGGDAPFCTRLNHSYTGMWAPERSAEARGNLTRPPGSGEDCGSVSSpecifications
| PTGER3 | |
| Polyclonal | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| prostaglandin E receptor 3 (subtype EP3) | |
| PTGER3 | |
| Recombinant protein corresponding to human PTGER3. | |
| RUO | |
| 5733 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| EP3/EP3-I/EP3-II/EP3-III/EP3-IV/EP3e/MGC141828/MGC141829/MGC27302 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction