missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGER3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24761-1-AP-20UL
This item is not returnable.
View return policy
Description
Receptor for prostaglandin E2 (PGE2); the EP3 receptor may be involved in inhibition of gastric acid secretion, modulation of neurotransmitter release in central and peripheral neurons, inhibition of sodium and water reabsorption in kidney tubulus and contraction in uterine smooth muscle. The activity of this receptor can couple to both the inhibition of adenylate cyclase mediated by G-I proteins, and to an elevation of intracellular calcium. The various isoforms have identical ligand binding properties but can interact with different second messenger systems.Specifications
| PTGER3 | |
| Polyclonal | |
| Unconjugated | |
| PTGER3 | |
| EP3, EP3 I, EP3 II, EP3 III, EP3 IV, EP3e, PGE receptor EP3 subtype, PGE2 R, PGE2 receptor EP3 subtype, Prostanoid EP3 receptor, PTGER3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19218, 5733 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P30557, P43115 | |
| PTGER3 | |
| PTGER3 Fusion Protein Ag21542 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction