missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGER3 Polyclonal Antibody

PTGER3 Polyclonal Antibody
Brand: Invitrogen PA534121
This item is not returnable.
View return policy
Description
PTGER3 Polyclonal Antibody for IHC (P)
Specifications
| PTGER3 | |
| Polyclonal | |
| Unconjugated | |
| PTGER3 | |
| EP3; Ep3 receptor; EP3A receptor; EP3B receptor; EP3C receptor; EP3D receptor; EP3e; EP3-I; EP3-II; EP3-III; EP3-IV; EP3-VI; MGC141828; MGC141829; MGC27302; PGE receptor EP3 subtype; PGE receptor, EP3 subtype; PGE2 receptor EP3 subtype; PGE2-R; Pgerep3; pr; prostaglandin E receotor EP3 subtype 3 isoform; prostaglandin E receptor 3; prostaglandin E receptor 3 (subtype EP3); prostaglandin E receptor EP3 subtype; prostaglandin E2 receptor EP3 subtype; prostaglandin receptor (PGE-2); prostanoid EP3 receptor; PTGER3; Ptgerep3; Rep3; rEP3a; rEP3b | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P43115 | |
| PTGER3 | |
| Synthetic 17 amino acid peptide from 1st cytoplasmic domain of human PTGER3 / EP3. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction