missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTGER3 Monoclonal Antibody (5F5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA112647
This item is not returnable.
View return policy
Description
A suggested positive control for this product is porcine brain lysate.
CD172a, also known as signal-regulatory protein alpha, is a receptor-type transmembrane glycoprotein expressed on cells of myeloid origin, including granulocytes, dendritic cells (DCs), macrophages, mast cells and haematopoietic stem cells. CD172a acts as a substrate for several activated tyrosine kinases, including EGFR, PDGFR, src and insulin receptor and is involved in the negative regulation of receptor tyrosine kinase-coupled signaling pathways. Ligand binding of CD172a to integrin-associated protein CD47, results in tyrosine kinase phosphorylation of immunoreceptor tyrosine-based inhibitory motifs (ITIMs) within the cytoplasmic region of CD172a, mediating the recruitment and activation of the tyrosine phosphatases SHP-1 and SHP-2. These then act as regulators of cellular function, through dephosphorylation of specific substrates. Ligation of CD172a with CD47 has been demonstrated in several regulatory processes, including the inhibition of host cell phagocytosis by macrophages and the bi-directional activation of T cells and DCs.
Specifications
| PTGER3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.08% sodium azide | |
| P34979, P34980, P43115 | |
| PTGER3 | |
| Recombinant human EP3 Receptor. | |
| 100 μg | |
| Primary | |
| Human, Rat, Bovine | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 5F5 | |
| Unconjugated | |
| PTGER3 | |
| EP3; Ep3 receptor; EP3A receptor; EP3B receptor; EP3C receptor; EP3D receptor; EP3e; EP3-I; EP3-II; EP3-III; EP3-IV; EP3-VI; MGC141828; MGC141829; MGC27302; PGE receptor EP3 subtype; PGE receptor, EP3 subtype; PGE2 receptor EP3 subtype; PGE2-R; Pgerep3; pr; prostaglandin E receotor EP3 subtype 3 isoform; prostaglandin E receptor 3; prostaglandin E receptor 3 (subtype EP3); prostaglandin E receptor EP3 subtype; prostaglandin E2 receptor EP3 subtype; prostaglandin receptor (PGE-2); prostanoid EP3 receptor; PTGER3; Ptgerep3; Rep3; rEP3a; rEP3b | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 24929, 282330, 5733 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction