missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGER2/3 Antibody (3F12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-66718
This item is not returnable.
View return policy
Description
PTGER2/3 Monoclonal specifically detects PTGER2/3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PTGER2/3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.08% Sodium Azide | |
| EP2, EP3, EP3-I, EP3-II, EP3-III, EP3-IV, PGE receptor EP2 subtype, PGE receptor, EP3 subtype, PGE2 receptor EP2 subtype, PGE2 receptor EP3 subtype, prostaglandin E receptor 2 (subtype EP2), 53kDa, prostaglandin E receptor 3 (subtype EP3), prostaglandin E2 receptor EP2 subtype, prostaglandin E2 receptor EP3 subtype, prostaglandin receptor (PGE-2), Prostanoid EP2 receptor, Prostanoid EP3 receptor | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant human PTGER2 and PTGER3 receptor protein and mouse myeloma cells. | |
| 100 μg | |
| 5732 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 3F12 | |
| Western Blot 5-10 ug/ml | |
| P43116 | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction