missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTGER1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599888
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PE2R1.
The protein encoded by this gene is a member of the G protein-coupled receptor family. This protein is one of four receptors identified for prostaglandin E2 (PGE2). Through a phosphatidylinositol-calcium second messenger system, G-Q proteins mediate this receptor's activity. Knockout studies in mice suggested a role of this receptor in mediating algesia and in regulation of blood pressure. Studies in mice also suggested that this gene may mediate adrenocorticotropic hormone response to bacterial endotoxin.
Specifications
| PTGER1 | |
| Polyclonal | |
| Unconjugated | |
| Ptger1 | |
| 42kDa; EP1; EP1 prostanoid receptor; PGE receptor EP1 subtype; PGE receptor, EP1 subtype; PGE2 receptor EP1 subtype; prostaglandin E receptor 1; prostaglandin E receptor 1 (subtype EP1); prostaglandin E receptor 1 (subtype EP1), 42kD; prostaglandin E receptor 1 (subtype EP1), 42kDa; prostaglandin E receptor 1, subtype EP1; prostaglandin E receptor EP1 subtype; prostaglandin E2 receptor EP1 subtype; Prostanoid EP1 receptor; PTGER1; Ptgerep1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25637, 5731 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P34995, P70597 | |
| Ptger1 | |
| A synthesized peptide derived from human PTGER1(Accession P34995), corresponding to amino acid residues L202-R252. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction