missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTGDR Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534113
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey, Marmoset (95%) Dog, Pig (85%) Panda, Bovine (80%).
The protein encoded by this gene is a G-protein-coupled receptor. It has been shown to function as a prostanoid DP receptor. The activity of this receptor is mainly mediated by G-S proteins that stimulate adenylate cyclase resulting in an elevation of intracellular cAMP and Ca2+. Knockout studies in mice suggest that the ligand of this receptor, prostaglandin D2 (PGD2), functions as a mast cell-derived mediator to trigger asthmatic responses.
Specifications
| PTGDR | |
| Polyclonal | |
| Unconjugated | |
| PTGDR | |
| AS1; ASRT1; DP; Dp receptor; DP1; MGC49004; PGD receptor; PGD2 receptor; prostaglandin D2 receptor; prostaglandin D2 receptor (DP); Prostanoid DP receptor; PTGDR; PTGDR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5729 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q13258 | |
| PTGDR | |
| Synthetic 20 amino acid peptide from 3rd extracellular domain of human Prostaglandin D2 Receptor. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction