Learn More
Fisherbrand™ PTFE Wing Stir Bars
Provides stable stirring action. The PTFE wing stir bars are effective in small tapered and round bottom test, centrifuge, and sample tubes.
Brand:  Fisherbrand™ 001.2201.1
Product Code. 10042853
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Stable, general-purpose magnetic stir bar
  • For effective stirring in small tapered and round bottom test, centrifuge, and sample tubes
  • Autoclavable