missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Tube

Product Code. 10127921 Shop All Bürkle Products
Change view
Click to view available options
Flow Rate:
270 mL/stroke
400 mL/stroke
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
10127921 400 mL/stroke Each
10042329 270 mL/stroke Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10127921 Supplier Buerkle™ Supplier No. 56061000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The PTFE Ultrapure Barrel Pump with Discharge Tube is suitable for the transfer of ultra-pure liquids and those that attack conventional materials such as PP or PVC. The stopcock prevents vapours from escaping, and stops dripping after transfer.

  • Barrel pump with rigid discharge tube with stopcock made of PTFE
  • The barrel pump is also suitable for transferring ultra-pure liquids
  • The stopcock prevents hazardous vapours from escaping, and stops dripping after transfer
  • Immersion tube Ø 32 mm
  • All valves are made of solid PTFE/PVDF
  • Top quality - 3 years' warranty
TRUSTED_SUSTAINABILITY

Specifications

Applications Ultra-Pure Liquid Transfer, chemically aggressive liquids
Flow Rate 400 mL/stroke
Includes Barrel Screw Joints
Opening Size (Metric) Tube diameter of 32 mm
Depth (Metric) 95 cm
Pump Tube Length 1.2 m
Resistance Properties Chemicals
Type Barrel Pump
Material PTFE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.