missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ PTFE U-Shaped Stirrer Shafts

Product Code. 10175910
Change view
Click to view available options
Length (Metric):
350 mm
450 mm
600 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10175910 350 mm Each
10104541 600 mm Each
10257451 450 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10175910 Supplier Bohlender™ Supplier No. C38402
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Diameter (Metric) Shaft 8 mm
For Use With (Application) Strong, tangential flow with high shear rate in the margin area, little sediments on the wall of the vessel
Length (Metric) 350 mm
Quantity 1 Each
Type U-shaped Stirrer Shaft
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.