Learn More
Kartell™ PTFE Tapered Stoppers
Description
- Excellent chemical resistance
- Can withstand high temperatures
- Self-lubricating
- Suitable for foodstuff
Specifications
Specifications
| Certifications/Compliance | DIN Standards 12242 |
| For Use With (Application) | Suitable for foodstuff |
| Quantity | 1/Pk. |
| Standard Taper Joint | NS 19/26 |
| Autoclavable | Autoclavable |
| Diameter (Metric) Bottom | 16.4 mm |
| Material | PTFE |
| Product Type | Tapered Stopper |