missing translation for 'onlineSavingsMsg'
Learn More

PTFE sheet, 1.6mm (0.063in) thick

Product Code. 15442197
Change view
Click to view available options
Quantity:
300 x 300 mm
Packaging:
Shrink-wrapped
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15442197 300 x 300 mm Shrink-wrapped Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15442197 Supplier Thermo Scientific Alfa Aesar Supplier No. 045194.HB
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9002-84-0 | (C2F4)n | NaN g/mol

PTFE sheet is used for the manufacture of high-temperature seals, insulators and bearings used in semiconductor, aerospace and chemical processing industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9002-84-0
Packaging Shrink-wrapped
Molecular Formula (C2F4)n
MDL Number MFCD00084455
Quantity 300 x 300 mm
Solubility Information Insoluble in water.
SMILES FC(F)(-*)C(F)(F)-*
Molecular Weight (g/mol) NaN
Chemical Name or Material PTFE sheet

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.