Learn More
Glindemann™ PTFE Sealing Rings for taper joints (laboratory glassware)
Product Code. 10125930
Encompass_Preferred
Change view
Click to view available options
Size:
NS 10
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 45
NS 7
Unit Size:
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10125930 NS 7 Pack of 50
10268410 NS 10 Pack of 50
10228460 NS 12 Pack of 50
10636521 NS 14 Pack of 50
10782131 NS 19 Pack of 50
10646521 NS 24 Pack of 50
10390001 NS 29 Pack of 50
10257790 NS 34 Pack of 50
10686901 NS 45 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10125930 Supplier Glindemann™ Supplier No. K7
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
For grease-free sealing of ground joints and plugs, extra tight and extra clean. The alternative to ground collar, grip collar sleeve and stopper made of PTFE.
  • Air leak rate <10-8 to 10-6 mbar l/s (high vacuum, overpressure).
  • Solvent leak rate in containers <0.1 to 0.03mg/d.
  • Temperature resistant from -200 to +300°C.
  • The elastic ring (0.07mm thick) adheres to the ground joint without a groove, is reusable and fits to ground joint clamps.
  • Other sizes on request.
Especificaciones
Material PTFE
Size NS 7
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.