Learn More
BRAND™ PTFE Seal
For use with the Transferpettor™ line, specially designed to handle viscous, foaming liquids and high vapor pressure.
Brand:  BRAND™ 701922
Code : 03
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10169990
  • £20.50 / Pack of 3
In Stock
to see stock.
Add to basket
Description
Description
  • With DE-M marking
  • Provided with mounting block
  • Provided in packs of 3
Specifications
Specifications
Conformity certified
50 μL
Mounting block
PTFE
For Transferpettor™ 50 µL capacity
3/Pk.