Learn More
DWK Life Sciences PTFE Key with Thread, with three part retaining device
For use with polished DURAN™ disposable taps.
Brand: DWK Life Sciences 286220604
Code : NEW
Additional Details : Weight : 0.01700kg
Description
Disposable PTFE valve plugs in combination with the polished DURAN™ disposable valve represent a grease-free alternative. The 3-part tap safety device is already included in the scope of delivery.
- Made of PTFE with thread and 3-part valve locking device
- For use with polished DURAN™ one-way taps
- Grease-free working
Specifications
4 mm | |
No | |
50/Pk. |
PTFE | |
14.5 NS | |
For use with polished DURAN™ one-way taps |