Learn More
Fisherbrand™ PTFE Conical Joint Clamps
Use as a safe solution to clamping glass joints. The PTFE conical joint clamps include an encapsulated steel spring.
Brand:  Fisherbrand™ 030.014.1
Product Code. 11563292
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
PTFE joint clamps are chemically inert and chemically resistant. Temperature resistant, maintaining strength up to 280°C (480°F). Manufactured from pure PTFE, with totally encapsulated steel springs. They can be adjusted by hand and fit standard glass joints.