missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Disposable Ground Joint Sleeves, PTFE
Description
- 50μm thickness
- Simplifies use of joint clips
- Fits like a second skin without adding bulk
Specifications
Specifications
| Material | PTFE |
| Product Type | Ground Joint Sleeve |
| Quantity | 10Pack |
| Temperature Range (Metric) | –200 to 260°C |
| Standard Taper Joint No. | NS 19/26 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?