missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCAT™ fittings are available in the following versions:
- PFA fitting with integrated ferrule in various colors and diameters
- Tube connectors in various diameters
Specifications
Specifications
| Type | Fitting |
| Color | White |
| Diameter (Metric) Outer | 4.76 mm |
| For Use With (Application) | Solvent Handling |
| For Use With | Safety Caps, Safety Waste Caps |
| Material | PTFE |
| Autoclavable | Autoclavable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?